This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07311231_m1
See other TIMD2 GE Assays ›
Gene Symbol:
Timd2
Gene Name:
T-cell immunoglobulin and mucin domain containing 2
Gene Aliases:
Tim2
Chromosome Location:
Chr.10: 31755473 - 31796423 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07311231_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Timd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013855.1 NP_001013877.1 3 - 4 428 97
XM_006246168.1 XP_006246230.1 4 - 5 713 97
XM_008767667.1 XP_008765889.1 4 - 5 476 97
XM_008767668.1 XP_008765890.1 4 - 5 476 97
XM_008767669.1 XP_008765891.1 4 - 5 968 97
GenBank mRNA BC089832.1 - 3 - 4 428 7324518 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Timd2

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0