This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06412847_s1
See other F2R GE Assays ›
Gene Symbol:
F2r
Gene Name:
coagulation factor II (thrombin) receptor
Gene Aliases:
Par1, TRGPC
Chromosome Location:
Chr.2: 26118760 - 26135340 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06412847_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F2r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012950.2 NP_037082.2 2 2523 70
GenBank mRNA BC085759.1 - 2 2523 7106133 70
CV116337.1 - 1 322 70
M81642.1 - 2 2532 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F2r

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
connective tissue replacement involved in inflammatory response wound healing
negative regulation of glomerular filtration
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
protein kinase C-activating G-protein coupled receptor signaling pathway
tyrosine phosphorylation of STAT protein
STAT protein import into nucleus
establishment of synaptic specificity at neuromuscular junction
blood coagulation
positive regulation of cell proliferation
negative regulation of cell proliferation
response to wounding
positive regulation of phosphatidylinositol 3-kinase signaling
obsolete death
platelet activation
regulation of blood coagulation
positive regulation of blood coagulation
positive regulation of cell migration
response to lipopolysaccharide
regulation of interleukin-1 beta production
positive regulation of collagen biosynthetic process
positive regulation of Rho protein signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
positive regulation of JAK-STAT cascade
homeostasis of number of cells within a tissue
release of sequestered calcium ion into cytosol
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of calcium ion transport
regulation of sensory perception of pain
positive regulation of ERK1 and ERK2 cascade
thrombin receptor signaling pathway
negative regulation of renin secretion into blood stream
positive regulation of interleukin-8 secretion
positive regulation of interleukin-6 secretion
G-protein alpha-subunit binding
G-protein coupled receptor activity
receptor binding
thrombin receptor activity
G-protein beta-subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0