This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06393323_m1
See other EDARADD GE Assays ›
Gene Symbol:
Edaradd
Gene Name:
EDAR-associated death domain
Gene Aliases:
RGD1564010
Chromosome Location:
Chr.17: 90802094 - 90843358 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06393323_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Edaradd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008771925.1 XP_008770147.1 4 - 5 754 83
XM_008771926.1 XP_008770148.1 6 - 7 794 83
XM_574055.6 XP_574055.4 5 - 6 906 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

hair follicle development
signal transduction
odontogenesis of dentin-containing tooth
trachea gland development

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0