This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06264521_s1
See other DIRAS3 GE Assays ›
Gene Symbol:
Diras3
Gene Name:
DIRAS family, GTP-binding RAS-like 3
Gene Aliases:
RGD1565168
Chromosome Location:
Chr.6: 135938345 - 135940205 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06264521_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Diras3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_345725.7 XP_345726.2 1 319 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_345725.7 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0