This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06239702_s1
See other LOC690911 GE Assays ›
Gene Symbol:
LOC690911
Gene Name:
similar to Msx2-interacting protein (SPEN homolog) (SMART/HDAC1-associated repressor protein)
Gene Aliases:
RGD1564662
Chromosome Location:
Chr.5: 160015573 - 160070915 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06239702_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC690911
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271495.1 NP_001258424.1 10 4380 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of neurogenesis
nucleotide binding
RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
single-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0