This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04419337_s1
See other CEBPB GE Assays ›
Gene Symbol:
Cebpb
Gene Name:
CCAAT/enhancer binding protein (C/EBP), beta
Gene Aliases:
Il6dbp, NF-IL6, TCF5
Chromosome Location:
Chr.3: 164424106 - 164425933 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04419337_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cebpb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301715.1 NP_001288644.1 1 720 58
NM_001301720.1 NP_001288649.1 1 720 58
NM_024125.5 NP_077039.3 1 720 58
GenBank mRNA BC129071.1 - 1 710 7458221 58
M57235.1 - 1 707 58
S77528.1 - 1 1105 58
X60769.1 - 1 439 58

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cebpb

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

embryonic placenta development
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
neuron differentiation
response to lipopolysaccharide
mammary gland epithelial cell proliferation
response to endoplasmic reticulum stress
negative regulation of neuron apoptotic process
regulation of interleukin-6 biosynthetic process
fat cell differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
mammary gland epithelial cell differentiation
regulation of transcription involved in cell fate commitment
cellular response to amino acid stimulus
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
glucocorticoid receptor binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300017753.rn.1 TC0300002372.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300017755.rn.1 TC0300002372.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0