This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244023_s1
See other MIR223 GE Assays ›
Gene Symbol:
Mir223
Gene Name:
microRNA 223
Gene Aliases:
rno-mir-223
Chromosome Location:
Chr.X: 65367812 - 65367921 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244023_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir223
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031936.1 1 51 108

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream serine/arginine repetitive matrix protein 4-like LOC102551035 -31.7kb Anti-sense Chr. X - 65335316 - 65336069 on NCBI Build Rnor_6.0
Downstream V-set and immunoglobulin domain containing 4 Vsig4 2.9kb Anti-sense Chr. X - 65370851 - 65400298 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

regulation of gene expression
positive regulation of granulocyte differentiation
gene silencing by miRNA
response to oxygen levels
cellular response to lipopolysaccharide
cellular response to estrogen stimulus
positive regulation of cell proliferation in bone marrow

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00003441.rn.1 TC0X00000553.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0