This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244019_s1
See other MIR217 GE Assays ›
Gene Symbol:
Mir217
Gene Name:
microRNA 217
Gene Aliases:
rno-mir-217
Chromosome Location:
Chr.14: 113119529 - 113119633 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244019_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir217
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031929.1 1 48 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 216a Mir216a rno-mir-216a -7.3kb Sense Chr. 14 - 113112129 - 113112234 on NCBI Build Rnor_6.0
Downstream EGF-containing fibulin-like extracellular matrix protein 1 Efemp1 T16 82.8kb Sense Chr. 14 - 113202386 - 113294993 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of gene expression
negative regulation of osteoblast differentiation
cellular response to lipopolysaccharide
cellular response to amino acid stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400008516.rn.1 TC1400001180.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0