This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04231454_s1
See other MIR139 GE Assays ›
Gene Symbol:
Mir139
Gene Name:
microRNA mir-139
Gene Aliases:
rno-mir-139
Chromosome Location:
Chr.1: 166589545 - 166589612 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04231454_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir139
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031886.1 1 40 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphodiesterase 2A,
cGMP-stimulated
Pde2a Pde2,
Pde2a2
Sense Chr. 1 - 166534643 - 166626158 on NCBI Build Rnor_6.0
Downstream ADP-ribosyltransferase 2b Art2b ARTC2,
Ag-F,
Art2,
Art2a,
LY2,
Ly-2,
PtaA,
RT6.1,
RT6.2,
Rt6
77.7kb Anti-sense Chr. 1 - 166667304 - 166685081 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100016830.rn.1 TC0100002168.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0