This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04224737_u1
See other F2RL3 GE Assays ›
Gene Symbol:
F2rl3
Gene Name:
coagulation factor II (thrombin) receptor-like 3
Gene Aliases:
PAR4
Chromosome Location:
Chr.16: 18817797 - 18819790 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04224737_u1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F2rl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053808.1 NP_446260.1 2 646 65
GenBank mRNA AF269246.2 - 2 646 65
AF310216.1 - 2 673 65

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F2rl3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
positive regulation of release of sequestered calcium ion into cytosol
thrombin receptor signaling pathway
G-protein coupled receptor activity
thrombin receptor activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P519917 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519917 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519917 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P519917 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P519917 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10940461 - -
Affymetrix Rat Gene 1.1 ST Array 10940461 - -
Rat Genome 230 2.0 Array 1369800_at - -
Rat Genome 230 2.0 Array 1369800_PM_at - -
Affymetrix HT Rat Focus Array 1369800_PM_at - -
Affymetrix HT Rat Focus Array 1369800_at - -
Rat Expression Set 230 1369800_at - -
Rat Expression Set 230 1369800_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10940461 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21380153 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0