This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04223737_u1
See other PALLD GE Assays ›
Gene Symbol:
Palld
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
-
Chromosome Location:
Chr.16: 31349138 - 31390340 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04223737_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Palld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751585.3 XP_003751633.1 2 1191 86
GenBank mRNA BC088409.1 - 2 1145 7189104 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600000411.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P762053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 561693 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857430 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 397905 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 695568 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 785056 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 507996 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 943476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 649505 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 279667 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 241956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 282059 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 153858 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 163484 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 59951 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 86995 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 135552 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 105363 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762053 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762053 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P762053 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P464083 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P174245 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P762053 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 69730 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 77155 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17952 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 51116 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 22592 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10940632 - -
Affymetrix Rat Gene 1.1 ST Array 10940632 - -
Rat Genome 230 2.0 Array 1384253_at - -
Rat Genome 230 2.0 Array 1384253_PM_at - -
Affymetrix HT Rat Focus Array 1384253_PM_at - -
Affymetrix HT Rat Focus Array 1384253_at - -
Affymetrix Rat Clariom S Assay PSR1600003862.rn.1 TC1600000411.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600003867.rn.1 TC1600000411.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600003861.rn.1 TC1600000411.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600003870.rn.1 TC1600000411.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600003869.rn.1 TC1600000411.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600003863.rn.1 TC1600000411.rn.1 RTA TC
Rat Expression Set 230 1384253_PM_at - -
Rat Expression Set 230 1384253_at - -
Affymetrix Rat Gene 1.0 ST Array 10940632 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0