This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02104920_s1
See other PROX2 GE Assays ›
Gene Symbol:
Prox2
Gene Name:
prospero homeobox 2
Gene Aliases:
RGD1310315
Chromosome Location:
Chr.6: 108911705 - 108920767 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02104920_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prox2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191771.1 NP_001178700.1 4 1647 83

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
multicellular organism development
DNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164245 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164245 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164245 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164245 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213380 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600020301.rn.1 TC0600002863.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600020303.rn.1 TC0600002863.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600020302.rn.1 TC0600002863.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600020306.rn.1 TC0600002863.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21359210 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1349927 - -
RatRef-12 Expression BeadChip ILMN_64099 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0