This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01771057_g1
See other GPR143 GE Assays ›
Gene Symbol:
Gpr143
Gene Name:
G protein-coupled receptor 143
Gene Aliases:
RGD1565799
Chromosome Location:
Chr.X: 23668363 - 23693162 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01771057_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106958.1 NP_001100428.1 7 - 8 948 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0X00000251.rn.1

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
melanosome localization
melanosome transport
melanosome organization
calcium-mediated signaling using intracellular calcium source
phosphatidylinositol-mediated signaling
regulation of calcium-mediated signaling
G-protein coupled receptor activity
dopamine binding
L-DOPA receptor activity
L-DOPA binding
tyrosine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00001901.rn.1 TC0X00000251.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0