This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01756911_gH
See other AGPAT6 GE Assays ›
Gene Symbol:
Agpat6
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 6
Gene Aliases:
RGD1310520
Chromosome Location:
Chr.16: 73615949 - 73649769 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01756911_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001047849.1 NP_001041314.1 9 - 10 1894 95
GenBank mRNA BC082092.1 - 6 - 7 740 95
BC161809.1 - 8 - 9 1180 8374387 95
DQ480750.1 - 9 - 10 1894 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600000858.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glandular epithelial cell maturation
fatty acid metabolic process
acyl-CoA metabolic process
triglyceride metabolic process
phosphatidylcholine biosynthetic process
lactation
lipid biosynthetic process
triglyceride biosynthetic process
mammary gland development
regulation of multicellular organism growth
diacylglycerol metabolic process
1-acylglycerol-3-phosphate O-acyltransferase activity
glycerol-3-phosphate O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600006606.rn.1 TC1600000858.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0