This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01749600_m1
See other AKIRIN1 GE Assays ›
Gene Symbol:
Akirin1
Gene Name:
akirin 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 141415226 - 141430627 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01749600_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Akirin1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030054.1 NP_001025225.1 4 - 5 660 142
GenBank mRNA CB761626.1 - 2 - 3 214 142

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500003556.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Akirin1

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16894 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 47462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853863 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 960274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 206502 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 855515 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 499293 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 782792 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 138975 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 414367 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 375681 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 75350 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 173677 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 155353 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16894 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16894 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16894 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16894 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 14375 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 73144 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10879670 - -
Affymetrix Rat Gene 1.0 ST Array 10879668 - -
Affymetrix Rat Gene 1.0 ST Array 10879669 - -
Affymetrix Rat Gene 1.0 ST Array 10879672 - -
Affymetrix Rat Gene 1.0 ST Array 10879671 - -
Affymetrix Rat Gene 1.1 ST Array 10879667 - -
Rat Genome 230 2.0 Array 1392908_PM_at - -
Rat Genome 230 2.0 Array 1392908_at - -
Affymetrix HT Rat Focus Array 1392908_PM_at - -
Affymetrix HT Rat Focus Array 1392908_at - -
Affymetrix Rat Clariom S Assay PSR0500026540.rn.1 TC0500003556.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500026541.rn.1 TC0500003556.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500026542.rn.1 TC0500003556.rn.1 RTA TC
Rat Expression Set 230 1392908_PM_at - -
Rat Expression Set 230 1392908_at - -
Affymetrix Rat Gene 1.0 ST Array 10879667 - -
Rat Genome U34 Set rc_AI070128_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21521796 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0