This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01648538_m1
See other PRSS30 GE Assays ›
Gene Symbol:
Prss30
Gene Name:
protease, serine, 30
Gene Aliases:
Tmprss8, tmsp-1
Chromosome Location:
Chr.10: 13348289 - 13355837 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01648538_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199371.1 NP_955403.1 4 - 5 535 87
XM_006245922.2 XP_006245984.1 5 - 6 726 87
XM_006245923.2 XP_006245985.1 5 - 6 734 87
XM_008767553.1 XP_008765775.1 5 - 6 754 87
GenBank mRNA AB073023.1 - 4 - 5 535 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000002374.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prss30

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
sodium ion transport
serine-type endopeptidase activity
sodium channel regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000020927.rn.1 TC1000002374.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0