This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01535673_m1
See other UQCC1 GE Assays ›
Gene Symbol:
Uqcc1
Gene Name:
ubiquinol-cytochrome c reductase complex assembly factor 1
Gene Aliases:
Uqcc
Chromosome Location:
Chr.3: 151384965 - 151474112 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01535673_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uqcc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109446.2 NP_001102916.1 9 - 10 772 65
XM_006235361.2 XP_006235423.1 9 - 10 846 65
GenBank mRNA BC107807.1 - 9 - 10 772 7321008 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300004560.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s)

mitochondrial respiratory chain complex III assembly
positive regulation of mitochondrial translation

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P130154 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130154 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130154 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P130154 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130154 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1372589_PM_at - -
Rat Genome 230 2.0 Array 1372589_at - -
Affymetrix HT Rat Focus Array 1372589_at - -
Affymetrix HT Rat Focus Array 1372589_PM_at - -
Affymetrix Rat Clariom S Assay PSR0300034986.rn.1 TC0300004560.rn.1 RTA TC
Rat Expression Set 230 1372589_PM_at - -
Rat Expression Set 230 1372589_at - -
Rat Genome U34 Set rc_AI177689_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22342284 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0