This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01523554_m1
See other LPHN2 GE Assays ›
Gene Symbol:
Lphn2
Gene Name:
latrophilin 2
Gene Aliases:
Cirl-2, Cirl2, Cl2ac
Chromosome Location:
Chr.2: 254398951 - 254532604 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01523554_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lphn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF081148.1 - 16 - 17 3425 79
AF081149.1 - 16 - 17 3425 79
AF081150.1 - 16 - 17 3425 79
AF081151.1 - 16 - 17 3425 79
AF081152.1 - 16 - 17 3425 79
AF081153.1 - 16 - 17 3425 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0