This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01515586_g1
See other TNFRSF14 GE Assays ›
Gene Symbol:
Tnfrsf14
Gene Name:
tumor necrosis factor receptor superfamily, member 14
Gene Aliases:
-
Chromosome Location:
Chr.5: 172328259 - 172337219 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01515586_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015034.1 NP_001015034.1 2 - 3 255 82
XM_006239586.2 XP_006239648.1 3 - 4 1140 82
XM_006239587.2 XP_006239649.1 3 - 4 1141 82
GenBank mRNA BC091202.1 - 2 - 3 255 7380378 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500004189.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf14

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine secretion involved in immune response
biological_process
negative regulation of alpha-beta T cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of T cell migration
molecular_function
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500031942.rn.1 TC0500004189.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0