This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01515505_m1
See other PHACTR4 GE Assays ›
Gene Symbol:
Phactr4
Gene Name:
phosphatase and actin regulator 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 150533895 - 150609422 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01515505_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phactr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008764195.1 XP_008762417.1 11 - 12 2273 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
neural tube closure
Rho protein signal transduction
actin cytoskeleton organization
positive regulation of catalytic activity
enteric nervous system development
regulation of cell cycle
closure of optic fissure
negative regulation of integrin-mediated signaling pathway
actin binding
protein phosphatase 1 binding
protein phosphatase type 1 activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0