This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01511891_m1
See other GPR116 GE Assays ›
Gene Symbol:
Gpr116
Gene Name:
G protein-coupled receptor 116
Gene Aliases:
Gprhep, Ig-Hepta
Chromosome Location:
Chr.9: 20091121 - 20195708 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01511891_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr116
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139110.1 NP_620810.1 20 - 21 4144 106
XM_006244594.2 XP_006244656.1 21 - 22 4479 106
XM_006244597.2 XP_006244659.1 20 - 21 4358 106
XM_008766832.1 XP_008765054.1 22 - 23 4547 106
XM_008766833.1 XP_008765055.1 22 - 23 4472 106
XM_008766835.1 XP_008765057.1 21 - 22 4426 106
GenBank mRNA AB019120.1 - 20 - 21 4144 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001656.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr116

Gene Ontology Categories:

Function(s) Process(es)

energy reserve metabolic process
G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
glucose homeostasis
surfactant homeostasis
fat cell differentiation
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P992535 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 874754 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 343405 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773170 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 73448 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 726271 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 784064 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 18817 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 409116 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171809 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 32687 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 259939 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 135905 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 144045 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 504892 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 929093 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 412132 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 775007 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 168031 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 879473 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 361118 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 837307 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120981 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 559817 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 322950 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 902980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 799282 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 966009 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 372002 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 118856 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 61707 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263700 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 878304 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 858708 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150136 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 890935 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 73350 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203641 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 61390 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 116777 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 22454 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 934043 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 447604 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 812440 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 622444 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 427945 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 630421 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 681956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 49320 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 38149 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 363393 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544393 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 297956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 768647 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263656 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894247 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577907 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 407240 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 304244 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 7807 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 223380 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 744092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 230123 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 777072 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 159544 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 621025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 64244 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 488880 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 167796 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 131692 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145568 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 90255 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 164406 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 179461 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 89704 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 48081 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 84671 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 58298 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 179764 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 108147 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 131542 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 46302 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 32787 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P992535 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P992535 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P992535 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P992535 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700914339 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80946 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 40586 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 5111 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 88420 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 67809 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 24399 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 88470 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80364 - -
Agilent Rat cDNA Microarray (G4105A) 700914339 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10926699 - -
Affymetrix Rat Gene 1.0 ST Array 10926718 - -
Affymetrix Rat Gene 1.0 ST Array 10926703 - -
Affymetrix Rat Gene 1.0 ST Array 10926711 - -
Affymetrix Rat Gene 1.0 ST Array 10926721 - -
Affymetrix Rat Gene 1.0 ST Array 10926715 - -
Affymetrix Rat Gene 1.0 ST Array 10926712 - -
Affymetrix Rat Gene 1.0 ST Array 10926704 - -
Affymetrix Rat Gene 1.0 ST Array 10926709 - -
Affymetrix Rat Gene 1.0 ST Array 10926701 - -
Affymetrix Rat Gene 1.0 ST Array 10926702 - -
Affymetrix Rat Gene 1.0 ST Array 10926716 - -
Affymetrix Rat Gene 1.0 ST Array 10926707 - -
Affymetrix Rat Gene 1.0 ST Array 10926722 - -
Affymetrix Rat Gene 1.0 ST Array 10926705 - -
Affymetrix Rat Gene 1.0 ST Array 10926713 - -
Affymetrix Rat Gene 1.0 ST Array 10926720 - -
Affymetrix Rat Gene 1.0 ST Array 10926700 - -
Affymetrix Rat Gene 1.0 ST Array 10926708 - -
Affymetrix Rat Gene 1.0 ST Array 10926714 - -
Affymetrix Rat Gene 1.0 ST Array 10926719 - -
Affymetrix Rat Gene 1.0 ST Array 10926717 - -
Affymetrix Rat Gene 1.0 ST Array 10926706 - -
Affymetrix Rat Gene 1.1 ST Array 10926698 - -
Rat Genome 230 2.0 Array 1398500_at - -
Rat Genome 230 2.0 Array 1376767_at - -
Rat Genome 230 2.0 Array 1388109_at - -
Rat Genome 230 2.0 Array 1376767_PM_at - -
Rat Genome 230 2.0 Array 1398500_PM_at - -
Rat Genome 230 2.0 Array 1388109_PM_at - -
Affymetrix HT Rat Focus Array 1376767_at - -
Affymetrix HT Rat Focus Array 1388109_at - -
Affymetrix HT Rat Focus Array 1388109_PM_at - -
Affymetrix HT Rat Focus Array 1376767_PM_at - -
Affymetrix Rat Clariom S Assay PSR0900013618.rn.1 TC0900001656.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900013619.rn.1 TC0900001656.rn.1 RTA TC
Rat Expression Set 230 1376767_at - -
Rat Expression Set 230 1388109_at - -
Rat Expression Set 230 1388109_PM_at - -
Rat Expression Set 230 1376767_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10926698 - -
Rat Genome U34 Set rc_AA900213_at - -
Rat Genome U34 Set rc_AA997889_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21347905 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1365711 - -
RatRef-12 Expression BeadChip ILMN_61931 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0