This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01507832_m1
See other CREBZF GE Assays ›
Gene Symbol:
Crebzf
Gene Name:
CREB/ATF bZIP transcription factor
Gene Aliases:
RGD1565584
Chromosome Location:
Chr.1: 156290215 - 156296891 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01507832_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crebzf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073439.1 1 - 2 1870 105
NR_073440.1 2 - 3 1659 105
NR_073441.1 2 - 3 1659 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006039.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
response to virus
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
regulation of sequence-specific DNA binding transcription factor activity
transcription factor activity, sequence-specific DNA binding
identical protein binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100047964.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047970.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047972.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047965.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047962.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047960.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047961.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047963.rn.1 TC0100006039.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047971.rn.1 TC0100006039.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0