This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01505452_g1
See other APOBEC3B GE Assays ›
Gene Symbol:
Apobec3b
Gene Name:
apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B
Gene Aliases:
Apobec3, Apobec3f
Chromosome Location:
Chr.7: 121108114 - 121128346 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01505452_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apobec3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033703.1 NP_001028875.1 6 - 7 1128 94
XM_008765715.1 XP_008763937.1 6 - 7 2211 94
XM_008765716.1 XP_008763938.1 7 - 8 2366 94
GenBank mRNA BC101878.1 - 6 - 7 1128 7929998 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700001464.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apobec3b

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
metabolic process
negative regulation of transposition
negative regulation of viral genome replication
innate immune response
defense response to virus
zinc ion binding
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
poly(A) RNA binding
deoxycytidine deaminase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700010977.rn.1 TC0700001464.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700010976.rn.1 TC0700001464.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700010978.rn.1 TC0700001464.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0