Assay Details
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106139.1 | NP_001099609.1 | 14 - 15 | 1951 | 117 | |
XM_006254859.2 | XP_006254921.1 | 12 - 13 | 1720 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132721 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21163 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132721 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132716 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132716 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132721 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132716 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132721 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P436240 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142573 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51277 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48345 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10802319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802315 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802321 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802316 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10802308 | - | - |
Rat Genome 230 2.0 Array | 1384970_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384970_at | - | - |
Affymetrix HT Rat Focus Array | 1384970_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384970_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800005137.rn.1 | TC1800000701.rn.1 | RTA TC |
Rat Expression Set 230 | 1384970_PM_at | - | - |
Rat Expression Set 230 | 1384970_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802308 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22156584 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372730 | - | - |
