This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01483810_m1
See other RNF2 GE Assays ›
Gene Symbol:
Rnf2
Gene Name:
ring finger protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 68829714 - 68859929 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01483810_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025667.1 NP_001020838.1 3 - 4 446 95
XM_006249990.2 XP_006250052.1 3 - 4 384 95
XM_006249991.2 XP_006250053.1 3 - 4 466 95
GenBank mRNA AB568264.1 - 2 - 3 250 95
BC098941.1 - 3 - 4 446 7456372 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001620.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnf2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
gastrulation with mouth forming second
transcription, DNA-templated
anterior/posterior axis specification
histone ubiquitination
spinal cord development
response to retinoic acid
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
molecular_function
chromatin binding
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity
RING-like zinc finger domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300011083.rn.1 TC1300001620.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0