This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01478872_m1
See other EPS8L2 GE Assays ›
Gene Symbol:
Eps8l2
Gene Name:
EPS8-like 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 214327992 - 214353466 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01478872_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eps8l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108508.1 NP_001101978.1 1 - 2 343 68
XM_006230562.1 XP_006230624.1 4 - 5 473 68
GenBank mRNA BC166805.1 - 4 - 5 253 7939004 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001108508.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
regulation of Rho protein signal transduction
positive regulation of ruffle assembly
actin binding
Rho guanyl-nucleotide exchange factor activity
Rac guanyl-nucleotide exchange factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100022981.rn.1 TC0100002896.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100022985.rn.1 TC0100002896.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100022984.rn.1 TC0100002896.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header