This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01476693_s1
See other CEBPA GE Assays ›
Gene Symbol:
Cebpa
Gene Name:
CCAAT/enhancer binding protein (C/EBP), alpha
Gene Aliases:
DBPCEP
Chromosome Location:
Chr.1: 91363492 - 91366164 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01476693_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cebpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287577.1 NP_001274506.1 1 2570 134
NM_001287578.1 NP_001274507.1 1 2570 134
NM_001287579.1 NP_001274508.1 1 2570 134
NM_012524.3 NP_036656.1 1 2570 134

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cebpa

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

urea cycle
negative regulation of transcription from RNA polymerase II promoter
liver development
embryonic placenta development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
acute-phase response
mitochondrion organization
response to nutrient
cholesterol metabolic process
negative regulation of cell proliferation
myeloid cell differentiation
cell differentiation
macrophage differentiation
lung development
granulocyte differentiation
organ regeneration
response to vitamin B2
regulation of cell proliferation
glucose homeostasis
fat cell differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase III promoter
cell maturation
inner ear development
white fat cell differentiation
brown fat cell differentiation
response to glucocorticoid
cellular response to lithium ion
cellular response to organic cyclic compound
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
protein domain specific binding
protein complex binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
HMG box domain binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21826638 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0