This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01476098_m1
See other ALDH16A1 GE Assays ›
Gene Symbol:
Aldh16a1
Gene Name:
aldehyde dehydrogenase 16 family, member A1
Gene Aliases:
-
Chromosome Location:
Chr.1: 101138237 - 101151320 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01476098_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh16a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033706.1 NP_001028878.1 16 - 17 2310 106
XM_006229099.2 XP_006229161.1 16 - 17 2383 106
XM_006229100.1 XP_006229162.1 15 - 16 2169 106
GenBank mRNA BC101860.1 - 16 - 17 2310 7936953 106
FQ220018.1 - 7 - 8 822 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100005380.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh16a1

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
aldehyde dehydrogenase (NAD) activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067320 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 115042 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 425442 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 721269 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 909456 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 30968 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 807081 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 190927 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 705797 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 755270 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 26026 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 96708 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067320 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067320 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067320 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212934 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 3990 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 2811 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10721655 - -
Affymetrix Rat Gene 1.0 ST Array 10721662 - -
Affymetrix Rat Gene 1.0 ST Array 10721660 - -
Affymetrix Rat Gene 1.0 ST Array 10721652 - -
Affymetrix Rat Gene 1.0 ST Array 10721650 - -
Affymetrix Rat Gene 1.0 ST Array 10721658 - -
Affymetrix Rat Gene 1.0 ST Array 10721651 - -
Affymetrix Rat Gene 1.0 ST Array 10721654 - -
Affymetrix Rat Gene 1.0 ST Array 10721661 - -
Affymetrix Rat Gene 1.0 ST Array 10721657 - -
Affymetrix Rat Gene 1.0 ST Array 10721664 - -
Affymetrix Rat Gene 1.0 ST Array 10721666 - -
Affymetrix Rat Gene 1.0 ST Array 10721663 - -
Affymetrix Rat Gene 1.0 ST Array 10721659 - -
Affymetrix Rat Gene 1.0 ST Array 10721665 - -
Affymetrix Rat Gene 1.0 ST Array 10721656 - -
Affymetrix Rat Gene 1.0 ST Array 10721653 - -
Affymetrix Rat Gene 1.1 ST Array 10721649 - -
Rat Genome 230 2.0 Array 1379228_PM_at - -
Rat Genome 230 2.0 Array 1373128_at - -
Rat Genome 230 2.0 Array 1393994_at - -
Rat Genome 230 2.0 Array 1374661_at - -
Rat Genome 230 2.0 Array 1379222_at - -
Rat Genome 230 2.0 Array 1393994_PM_at - -
Rat Genome 230 2.0 Array 1373128_PM_at - -
Rat Genome 230 2.0 Array 1379222_PM_at - -
Rat Genome 230 2.0 Array 1374661_PM_at - -
Rat Genome 230 2.0 Array 1379228_at - -
Affymetrix HT Rat Focus Array 1374661_at - -
Affymetrix HT Rat Focus Array 1374661_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100043732.rn.1 TC0100005380.rn.1 RTA TC
Rat Expression Set 230 1373128_PM_at - -
Rat Expression Set 230 1374661_at - -
Rat Expression Set 230 1374661_PM_at - -
Rat Expression Set 230 1373128_at - -
Affymetrix Rat Gene 1.0 ST Array 10721649 - -
Rat Genome U34 Set rc_AA875665_g_at - -
Rat Genome U34 Set rc_AA875665_at - -
Rat Genome U34 Set rc_AI029499_at - -
Rat Genome U34 Set rc_AI008125_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21060254 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1353249 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0