This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01475838_m1
See other LOC102550682 GE Assays ›
Gene Symbol:
LOC102550682
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 5-like
Gene Aliases:
-
Chromosome Location:
Chr.1: 79289038 - 79358187 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01475838_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102550682
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006228530.1 XP_006228592.1 8 - 9 2702 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21073633 - -
ABI Rat Genome Survey Microarray 21968891 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0