Assay Details
Gene Symbol: Col8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107100.1 | NP_001100570.1 | 3 - 4 | 523 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P140684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13012 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140684 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140684 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P140684 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P140684 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95314 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70542 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22116 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10750686 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750687 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10750685 | - | - |
Rat Genome 230 2.0 Array | 1393891_at | - | - |
Rat Genome 230 2.0 Array | 1393891_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393891_at | - | - |
Affymetrix HT Rat Focus Array | 1393891_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100002171.rn.1 | TC1100000453.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100002169.rn.1 | TC1100000453.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100002163.rn.1 | TC1100000453.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100002164.rn.1 | TC1100000453.rn.1 | RTA TC |
Rat Expression Set 230 | 1393891_PM_at | - | - |
Rat Expression Set 230 | 1393891_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750685 | - | - |
Rat Genome U34 Set | S63458_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21022328 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362033 | - | - |
