This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01474619_m1
See other HCLS1 GE Assays ›
Gene Symbol:
Hcls1
Gene Name:
hematopoietic cell specific Lyn substrate 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 66736966 - 66759402 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01474619_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hcls1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011898.1 NP_001011898.1 7 - 8 597 84
GenBank mRNA BC079081.1 - 7 - 8 597 7122756 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hcls1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
response to hormone
positive regulation of phosphatidylinositol 3-kinase signaling
actin filament polymerization
erythrocyte differentiation
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of tyrosine phosphorylation of STAT protein
positive regulation of transcription factor import into nucleus
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
negative regulation of leukocyte apoptotic process
RNA polymerase II transcription factor binding
actin binding
SH3 domain binding
protein kinase binding
protein complex binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 757917 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826804 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 72119 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 936170 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 431216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 503541 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 405891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 225546 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 402253 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 566255 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 332628 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 838554 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 841655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 130092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 66519 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396154 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 491830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 973399 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 458123 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 126571 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 105146 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15879 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134741 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134741 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134741 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21641 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21322 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 74928 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 25972 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10754303 - -
Affymetrix Rat Gene 1.0 ST Array 10754298 - -
Affymetrix Rat Gene 1.0 ST Array 10754308 - -
Affymetrix Rat Gene 1.0 ST Array 10754305 - -
Affymetrix Rat Gene 1.0 ST Array 10754310 - -
Affymetrix Rat Gene 1.0 ST Array 10754306 - -
Affymetrix Rat Gene 1.0 ST Array 10754309 - -
Affymetrix Rat Gene 1.0 ST Array 10754300 - -
Affymetrix Rat Gene 1.0 ST Array 10754297 - -
Affymetrix Rat Gene 1.0 ST Array 10754302 - -
Affymetrix Rat Gene 1.0 ST Array 10754307 - -
Affymetrix Rat Gene 1.0 ST Array 10754301 - -
Affymetrix Rat Gene 1.0 ST Array 10754299 - -
Affymetrix Rat Gene 1.0 ST Array 10754304 - -
Affymetrix Rat Gene 1.1 ST Array 10754296 - -
Rat Genome 230 2.0 Array 1380521_PM_at - -
Rat Genome 230 2.0 Array 1380521_at - -
Affymetrix HT Rat Focus Array 1380521_PM_at - -
Affymetrix HT Rat Focus Array 1380521_at - -
Affymetrix Rat Clariom S Assay PSR1100010210.rn.1 TC1100001677.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100010211.rn.1 TC1100001677.rn.1 RTA TC
Rat Expression Set 230 1380521_PM_at - -
Rat Expression Set 230 1380521_at - -
Affymetrix Rat Gene 1.0 ST Array 10754296 - -
Rat Genome U34 Set rc_AA925692_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22272555 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1376482 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0