This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01474549_m1
See other LOC102554620 GE Assays ›
Gene Symbol:
LOC102554620 Vwce
Gene Name:
von Willebrand factor C and EGF domain-containing protein-like
von Willebrand factor C and EGF domains
Gene Aliases:
-
RGD1309910
Chromosome Location:
Chr.Un|NW_007906915: 65166 - 86730 on Build Rnor_6.0
Chromosome Location:
Chr.1: 226687258 - 226717989 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01474549_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102554620
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008773895.1 XP_008772117.1 544 133
XM_008773896.1 XP_008772118.1 5 - 6 544 133
XM_008773897.1 XP_008772119.1 1011 133
XM_008773898.1 XP_008772120.1 5 - 6 544 133
Gene Symbol: Vwce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271311.1 NP_001258240.1 6 - 7 658 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003205.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell proliferation
regulation of signal transduction
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100026513.rn.1 TC0100003205.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header