This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01471491_m1
See other ZPBP2 GE Assays ›
Gene Symbol:
Zpbp2
Gene Name:
zona pellucida binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 86513654 - 86523375 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01471491_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zpbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007011.1 NP_001007012.1 5 - 6 725 61
XM_006247384.2 XP_006247446.1 5 - 6 1855 61
XM_006247385.2 XP_006247447.1 5 - 6 1782 61
GenBank mRNA AY251605.1 - 5 - 6 717 61
AY251606.1 - 4 - 5 651 61
BC082007.1 - 5 - 6 725 7131668 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000001579.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zpbp2

Gene Ontology Categories:

Function(s)

acrosome assembly
binding of sperm to zona pellucida

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000014014.rn.1 TC1000001579.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0