This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01461644_m1
See other HAVCR2 GE Assays ›
Gene Symbol:
Havcr2
Gene Name:
hepatitis A virus cellular receptor 2
Gene Aliases:
tim3
Chromosome Location:
Chr.10: 31561895 - 31589878 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01461644_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Havcr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100762.1 NP_001094232.1 6 - 7 756 132
XM_006246171.2 XP_006246233.1 5 - 6 668 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000000450.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037080 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 890578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 159081 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 108049 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 408848 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 132864 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 692336 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 904720 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 321260 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 405323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 552477 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 211766 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826248 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 804987 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 151872 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773935 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687164 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 334695 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 775433 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 602916 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140583 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 66328 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 139473 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 125670 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037080 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037080 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P037080 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543329 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546419 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 57717 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10733012 - -
Affymetrix Rat Gene 1.0 ST Array 10733011 - -
Affymetrix Rat Gene 1.0 ST Array 10733014 - -
Affymetrix Rat Gene 1.0 ST Array 10733009 - -
Affymetrix Rat Gene 1.0 ST Array 10733013 - -
Affymetrix Rat Gene 1.0 ST Array 10733008 - -
Affymetrix Rat Gene 1.0 ST Array 10733010 - -
Affymetrix Rat Gene 1.1 ST Array 10733007 - -
Affymetrix Rat Clariom S Assay PSR1000003833.rn.1 TC1000000450.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000003834.rn.1 TC1000000450.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10733007 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21906201 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356543 - -
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359061 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0