This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01459910_m1
See other KCTD2 GE Assays ›
Gene Symbol:
Kctd2
Gene Name:
potassium channel tetramerization domain containing 2
Gene Aliases:
RGD1566063
Chromosome Location:
Chr.10: 103972750 - 103984398 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01459910_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,681.00
Your Price
Online offer:
2681.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kctd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008768411.1 XP_008766633.1 3 - 4 578 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein homooligomerization
protein complex binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255954 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255954 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P255954 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255954 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.1 ST Array 10739543 - -
Rat Genome 230 2.0 Array 1389988_at - -
Rat Genome 230 2.0 Array 1389988_PM_at - -
Affymetrix HT Rat Focus Array 1389988_at - -
Affymetrix HT Rat Focus Array 1389988_PM_at - -
Rat Expression Set 230 1389988_PM_at - -
Rat Expression Set 230 1389988_at - -
Rat Genome U34 Set rc_AI030494_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21016760 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372689 - -
RatRef-12 Expression BeadChip ILMN_58413 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0