This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01451066_m1
See other ADRBK1 GE Assays ›
Gene Symbol:
Adrbk1
Gene Name:
adrenergic, beta, receptor kinase 1
Gene Aliases:
BARK1, GRK-2, Grk2
Chromosome Location:
Chr.1: 219536220 - 219544329 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01451066_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adrbk1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012776.1 NP_036908.1 14 - 15 1473 85
GenBank mRNA M87854.1 - 14 - 15 1473 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adrbk1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
desensitization of G-protein coupled receptor protein signaling pathway
negative regulation of the force of heart contraction by chemical signal
protein phosphorylation
response to oxidative stress
G-protein coupled receptor signaling pathway
tachykinin receptor signaling pathway
heart development
aging
response to organic cyclic compound
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
receptor internalization
positive regulation of catecholamine secretion
termination of G-protein coupled receptor signaling pathway
response to hydrogen peroxide
follicle-stimulating hormone signaling pathway
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of striated muscle contraction
cardiac muscle contraction
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein binding
ATP binding
Edg-2 lysophosphatidic acid receptor binding
beta-adrenergic receptor kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0