This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01450825_m1
See other EHD1 GE Assays ›
Gene Symbol:
Ehd1
Gene Name:
EH-domain containing 1
Gene Aliases:
RGD1306960
Chromosome Location:
Chr.1: 221644867 - 221667242 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01450825_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ehd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011939.2 NP_001011939.1 1 - 2 475 139
GenBank mRNA BC082030.1 - 1 - 2 429 139
BC160908.1 - 1 - 2 476 139
CV120970.1 - 1 - 2 475 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003101.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
endocytosis
positive regulation of cholesterol storage
endosomal transport
neuron projection development
endocytic recycling
low-density lipoprotein particle clearance
cholesterol homeostasis
protein homooligomerization
positive regulation of myoblast fusion
cellular response to nerve growth factor stimulus
positive regulation of endocytic recycling
calcium ion binding
protein binding
ATP binding
GTP binding
Rab GTPase binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P696669 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164797 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 783782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 778322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 390889 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 947834 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716733 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 479294 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687056 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323005 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 266864 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 307794 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 786842 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 145693 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 101385 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 546348 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 450166 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 834798 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 691904 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723547 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 154346 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 18834 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 60329 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164797 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P696669 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164797 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P696669 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P696669 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164797 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16917 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P696669 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 104104 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 61842 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10713274 - -
Affymetrix Rat Gene 1.0 ST Array 10713276 - -
Affymetrix Rat Gene 1.0 ST Array 10713275 - -
Affymetrix Rat Gene 1.0 ST Array 10713271 - -
Affymetrix Rat Gene 1.0 ST Array 10713272 - -
Affymetrix Rat Gene 1.0 ST Array 10713277 - -
Affymetrix Rat Gene 1.0 ST Array 10713273 - -
Affymetrix Rat Gene 1.1 ST Array 10713222 - -
Rat Genome 230 2.0 Array 1388623_at - -
Rat Genome 230 2.0 Array 1388623_PM_at - -
Rat Genome 230 2.0 Array 1372317_PM_at - -
Rat Genome 230 2.0 Array 1372317_at - -
Affymetrix HT Rat Focus Array 1372317_PM_at - -
Affymetrix HT Rat Focus Array 1388623_PM_at - -
Affymetrix HT Rat Focus Array 1372317_at - -
Affymetrix HT Rat Focus Array 1388623_at - -
Affymetrix Rat Clariom S Assay PSR0100025205.rn.1 TC0100003101.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100025206.rn.1 TC0100003101.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100025204.rn.1 TC0100003101.rn.1 RTA TC
Rat Expression Set 230 1372317_at - -
Rat Expression Set 230 1372317_PM_at - -
Rat Expression Set 230 1388623_at - -
Rat Expression Set 230 1388623_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10713222 - -
Rat Genome U34 Set rc_AI045370_at - -
Rat Genome U34 Set rc_AI059096_at - -
Rat Genome U34 Set rc_AI177867_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21257727 - -
ABI Rat Genome Survey Microarray 22095403 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1376189 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0