This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01432888_s1
See other FOXC2 GE Assays ›
Gene Symbol:
Foxc2
Gene Name:
forkhead box C2
Gene Aliases:
Fkh14, Fkhl14, Hfhbf3
Chromosome Location:
Chr.19: 53044379 - 53047081 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01432888_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Foxc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101680.1 NP_001095150.1 1 2633 118

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
ossification
blood vessel development
patterning of blood vessels
metanephros development
ureteric bud development
somitogenesis
kidney development
lymph vessel development
lymphangiogenesis
blood vessel remodeling
heart morphogenesis
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
Notch signaling pathway
heart development
cell proliferation
insulin receptor signaling pathway
response to hormone
positive regulation of endothelial cell migration
neural crest cell development
collagen fibril organization
positive regulation of cell adhesion mediated by integrin
embryonic heart tube development
positive regulation of vascular wound healing
camera-type eye development
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of organ growth
vascular endothelial growth factor receptor signaling pathway
paraxial mesoderm formation
paraxial mesodermal cell fate commitment
embryonic cranial skeleton morphogenesis
embryonic viscerocranium morphogenesis
embryonic skeletal system morphogenesis
artery morphogenesis
regulation of blood vessel size
ventricular cardiac muscle tissue morphogenesis
cardiac muscle cell proliferation
glomerular endothelium development
glomerular visceral epithelial cell differentiation
glomerular mesangial cell development
positive regulation of cell migration involved in sprouting angiogenesis
negative regulation of apoptotic process involved in outflow tract morphogenesis
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
chromatin DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P671089 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P671089 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P671089 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P671089 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P671089 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P633261 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1375607_PM_at - -
Rat Genome 230 2.0 Array 1377487_at - -
Rat Genome 230 2.0 Array 1377487_PM_at - -
Rat Genome 230 2.0 Array 1370795_PM_at - -
Rat Genome 230 2.0 Array 1370795_at - -
Rat Genome 230 2.0 Array 1375607_at - -
Affymetrix HT Rat Focus Array 1375607_at - -
Affymetrix HT Rat Focus Array 1370795_at - -
Affymetrix HT Rat Focus Array 1370795_PM_at - -
Affymetrix HT Rat Focus Array 1375607_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900006062.rn.1 TC1900000731.rn.1 RTA TC
Rat Expression Set 230 1377487_PM_at - -
Rat Expression Set 230 1370795_PM_at - -
Rat Expression Set 230 1375607_PM_at - -
Rat Expression Set 230 1370795_at - -
Rat Expression Set 230 1375607_at - -
Rat Expression Set 230 1377487_at - -
Rat Genome U34 Set L13193_at - -
Rat Genome U34 Set rc_AI059526_at - -
Rat Genome U34 Set rc_AI112837_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22360811 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0