This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01430793_m1
See other USP11 GE Assays ›
Gene Symbol:
Usp11
Gene Name:
ubiquitin specific peptidase 11
Gene Aliases:
Uhx1
Chromosome Location:
Chr.X: 1687820 - 1703991 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01430793_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,681.00
Your Price
Online offer:
2681.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Usp11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008861.2 NP_001008861.2 13 - 14 1770 89
GenBank mRNA BC090333.1 - 13 - 14 1770 7190710 89
BC105613.1 - 9 - 10 1183 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0X00001378.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Usp11

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P261266 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 60 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 156517 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 632250 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 499875 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 852250 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 836657 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 242910 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 778064 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 666901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 443648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 712471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 823758 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 301104 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 464319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 631187 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 123230 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 48152 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P261266 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P261266 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P261266 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P261266 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 65340 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 46646 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 12430 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10936549 - -
Affymetrix Rat Gene 1.0 ST Array 10936544 - -
Affymetrix Rat Gene 1.0 ST Array 10936557 - -
Affymetrix Rat Gene 1.0 ST Array 10936555 - -
Affymetrix Rat Gene 1.0 ST Array 10936540 - -
Affymetrix Rat Gene 1.0 ST Array 10936542 - -
Affymetrix Rat Gene 1.0 ST Array 10936560 - -
Affymetrix Rat Gene 1.0 ST Array 10936541 - -
Affymetrix Rat Gene 1.0 ST Array 10936552 - -
Affymetrix Rat Gene 1.0 ST Array 10936558 - -
Affymetrix Rat Gene 1.0 ST Array 10936556 - -
Affymetrix Rat Gene 1.0 ST Array 10936553 - -
Affymetrix Rat Gene 1.0 ST Array 10936550 - -
Affymetrix Rat Gene 1.0 ST Array 10936551 - -
Affymetrix Rat Gene 1.0 ST Array 10936543 - -
Affymetrix Rat Gene 1.0 ST Array 10936545 - -
Affymetrix Rat Gene 1.0 ST Array 10936554 - -
Affymetrix Rat Gene 1.0 ST Array 10936547 - -
Affymetrix Rat Gene 1.0 ST Array 10936548 - -
Affymetrix Rat Gene 1.0 ST Array 10936546 - -
Affymetrix Rat Gene 1.0 ST Array 10936559 - -
Affymetrix Rat Gene 1.1 ST Array 10936539 - -
Rat Genome 230 2.0 Array 1391938_at - -
Rat Genome 230 2.0 Array 1376171_PM_at - -
Rat Genome 230 2.0 Array 1391938_PM_at - -
Rat Genome 230 2.0 Array 1376171_at - -
Affymetrix HT Rat Focus Array 1376171_at - -
Affymetrix HT Rat Focus Array 1391938_at - -
Affymetrix HT Rat Focus Array 1391938_PM_at - -
Affymetrix HT Rat Focus Array 1376171_PM_at - -
Affymetrix Rat Clariom S Assay PSR0X00008628.rn.1 TC0X00001378.rn.1 RTA TC
Rat Expression Set 230 1376171_at - -
Rat Expression Set 230 1376171_PM_at - -
Rat Expression Set 230 1391938_PM_at - -
Rat Expression Set 230 1391938_at - -
Affymetrix Rat Gene 1.0 ST Array 10936539 - -
Rat Genome U34 Set rc_AA851303_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21376050 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1358251 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0