This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01418475_mH
See other CHRNA5 GE Assays ›
Gene Symbol:
Chrna5
Gene Name:
cholinergic receptor, nicotinic, alpha 5 (neuronal)
Gene Aliases:
-
Chromosome Location:
Chr.8: 59561817 - 59590172 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01418475_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chrna5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017078.2 NP_058774.2 3 - 4 359 136
GenBank mRNA AY574255.1 - 3 - 4 257 136
J05231.1 - 3 - 4 358 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000745.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chrna5

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

cation transport
chemical synaptic transmission
response to nicotine
behavioral response to nicotine
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
acetylcholine receptor activity
acetylcholine binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800005001.rn.1 TC0800000745.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0