This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01413444_m1
See other SRP72 GE Assays ›
Gene Symbol:
Srp72
Gene Name:
signal recognition particle 72
Gene Aliases:
-
Chromosome Location:
Chr.14: 33536123 - 33561141 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01413444_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srp72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170601.1 NP_001164072.1 1516 117
XM_008770133.1 XP_008768355.1 1962 117
GenBank mRNA BC107914.1 - 666 117
BC158784.1 - 264 7302022 117

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400011952.rn.1 TC1400001631.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400011943.rn.1 TC1400001631.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400011950.rn.1 TC1400001631.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400011951.rn.1 TC1400001631.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400011948.rn.1 TC1400001631.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0