This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01411797_g1
See other MPZ GE Assays ›
Gene Symbol:
Mpz
Gene Name:
myelin protein zero
Gene Aliases:
MPP, P0
Chromosome Location:
Chr.13: 89524349 - 89529248 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01411797_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017027.1 NP_058723.1 5 - 6 679 122
GenBank mRNA BC078859.1 - 679 122
K03242.1 - 5 - 6 679 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300000751.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mpz

Panther Classification:

Molecular Function -

myelin protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
cell-cell junction maintenance
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127576 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036965 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036972 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 41228 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 742136 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 558736 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 162878 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036965 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127576 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036970 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036970 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036965 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127576 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036965 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127576 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036970 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11783 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 46549 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10765516 - -
Affymetrix Rat Gene 1.0 ST Array 10765509 - -
Affymetrix Rat Gene 1.0 ST Array 10765515 - -
Affymetrix Rat Gene 1.0 ST Array 10765512 - -
Affymetrix Rat Gene 1.0 ST Array 10765514 - -
Affymetrix Rat Gene 1.0 ST Array 10765513 - -
Affymetrix Rat Gene 1.0 ST Array 10765510 - -
Affymetrix Rat Gene 1.0 ST Array 10765511 - -
Affymetrix Rat Gene 1.1 ST Array 10765508 - -
Rat Genome 230 2.0 Array 1368815_at - -
Rat Genome 230 2.0 Array 1368816_PM_at - -
Rat Genome 230 2.0 Array 1368815_PM_at - -
Rat Genome 230 2.0 Array 1368816_at - -
Affymetrix HT Rat Focus Array 1368815_PM_at - -
Affymetrix HT Rat Focus Array 1368816_at - -
Affymetrix HT Rat Focus Array 1368815_at - -
Affymetrix HT Rat Focus Array 1368816_PM_at - -
Affymetrix Rat Clariom S Assay PSR1300005183.rn.1 TC1300000751.rn.1 RTA TC
Rat Expression Set 230 1368815_PM_at - -
Rat Expression Set 230 1368815_at - -
Rat Expression Set 230 1368816_at - -
Rat Expression Set 230 1368816_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10765508 - -
Rat Genome U34 Set K03242_at - -
Rat Genome U34 Set rc_AA956483_at - -
Rat Genome U34 Set K03242_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21514660 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359421 - -
RatRef-12 Expression BeadChip ILMN_52927 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0