This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01408666_m1
See other EPB4.1L5 GE Assays ›
Gene Symbol:
Epb4.1l5
Gene Name:
erythrocyte membrane protein band 4.1-like 5
Gene Aliases:
Epb41l5
Chromosome Location:
Chr.13: 35622472 - 35673449 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01408666_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012023.1 NP_001012023.1 12 - 13 1178 95
GenBank mRNA BC090012.1 - 12 - 13 1178 7324591 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001260.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1l5

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis involved in differentiation
in utero embryonic development
somitogenesis
epithelial to mesenchymal transition
neural plate morphogenesis
positive regulation of cell-matrix adhesion
epithelial cell morphogenesis
apical constriction
substrate-dependent cell migration, cell attachment to substrate
ectoderm development
endoderm development
mesoderm development
mesoderm migration involved in gastrulation
unidimensional cell growth
posttranscriptional regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-cell adhesion
actin cytoskeleton organization
actomyosin structure organization
negative regulation of protein binding
positive regulation of protein binding
somite rostral/caudal axis specification
axial mesoderm development
axial mesoderm morphogenesis
paraxial mesoderm development
embryonic foregut morphogenesis
positive regulation of focal adhesion assembly
regulation of establishment of protein localization
left/right axis specification
cellular response to transforming growth factor beta stimulus
cytoskeletal protein binding
protein domain specific binding

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014371 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120783 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 219564 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 42833 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577570 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 898313 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 598533 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 257169 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 424476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 842643 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 80946 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 598573 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 113154 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 650078 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 898594 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203914 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 561796 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 20864 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 759571 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 445025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 23327 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 525281 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740630 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 524791 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 850629 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 563374 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 778395 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 854245 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 362427 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655329 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 757828 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 863127 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 830215 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 688274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 20320 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 212530 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 159101 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 109230 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 49673 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 80298 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 83703 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 171285 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014371 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014371 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014371 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P715347 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 57320 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 103801 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80341 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 39571 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10767129 - -
Affymetrix Rat Gene 1.0 ST Array 10767119 - -
Affymetrix Rat Gene 1.0 ST Array 10767124 - -
Affymetrix Rat Gene 1.0 ST Array 10767127 - -
Affymetrix Rat Gene 1.0 ST Array 10767126 - -
Affymetrix Rat Gene 1.0 ST Array 10767115 - -
Affymetrix Rat Gene 1.0 ST Array 10767122 - -
Affymetrix Rat Gene 1.0 ST Array 10767112 - -
Affymetrix Rat Gene 1.0 ST Array 10767128 - -
Affymetrix Rat Gene 1.0 ST Array 10767120 - -
Affymetrix Rat Gene 1.0 ST Array 10767125 - -
Affymetrix Rat Gene 1.0 ST Array 10767117 - -
Affymetrix Rat Gene 1.0 ST Array 10767113 - -
Affymetrix Rat Gene 1.0 ST Array 10767118 - -
Affymetrix Rat Gene 1.0 ST Array 10767116 - -
Affymetrix Rat Gene 1.0 ST Array 10767114 - -
Affymetrix Rat Gene 1.0 ST Array 10767121 - -
Affymetrix Rat Gene 1.0 ST Array 10767123 - -
Affymetrix Rat Gene 1.1 ST Array 10767102 - -
Rat Genome 230 2.0 Array 1374984_at - -
Rat Genome 230 2.0 Array 1374984_PM_at - -
Affymetrix HT Rat Focus Array 1374984_PM_at - -
Affymetrix HT Rat Focus Array 1374984_at - -
Affymetrix Rat Clariom S Assay PSR1300008324.rn.1 TC1300001260.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300008325.rn.1 TC1300001260.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300008336.rn.1 TC1300001260.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300008323.rn.1 TC1300001260.rn.1 RTA TC
Rat Expression Set 230 1374984_at - -
Rat Expression Set 230 1374984_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10767102 - -
Rat Genome U34 Set rc_AA858925_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0