This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01402912_g1
See other NPY1R GE Assays ›
Gene Symbol:
Npy1r
Gene Name:
neuropeptide Y receptor Y1
Gene Aliases:
NPY-1
Chromosome Location:
Chr.16: 24779480 - 24788985 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01402912_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npy1r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113357.1 NP_001106828.1 2 - 3 858 109
XM_006253024.2 XP_006253086.1 3 - 4 1253 109
XM_006253025.2 XP_006253087.1 3 - 4 1166 109
GenBank mRNA Z11504.1 - 1 - 2 753 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600001468.rn.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
glucose metabolic process
neuropeptide signaling pathway
chemical synaptic transmission
locomotory behavior
feeding behavior
regulation of blood pressure
sensory perception of pain
peristalsis
response to estradiol
regulation of multicellular organism growth
positive regulation of vasoconstriction
negative regulation of hormone secretion
negative regulation of cytosolic calcium ion concentration
peptide YY receptor activity
pancreatic polypeptide receptor activity
molecular_function
neuropeptide Y receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600011397.rn.1 TC1600001468.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600011398.rn.1 TC1600001468.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm