This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00821569_g1
See other RPS15A GE Assays ›
Gene Symbol:
Rps15a
Gene Name:
ribosomal protein S15a
Gene Aliases:
-
Chromosome Location:
Chr.1: 187759855 - 187766734 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00821569_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps15a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053982.1 NP_446434.1 2 - 3 216 96
XM_006230097.1 XP_006230159.1 3 - 4 263 96
XM_006230098.1 XP_006230160.1 3 - 4 306 96
GenBank mRNA BC058452.1 - 3 - 4 241 6921126 96
FQ210209.1 - 3 - 4 96
FQ211215.1 - 3 - 4 241 96
FQ212042.1 - 3 - 4 237 96
FQ212257.1 - 3 - 4 235 96
FQ214481.1 - 3 - 4 237 96
FQ220418.1 - 1 - 2 61 96
FQ220927.1 - 3 - 4 233 96
FQ220997.1 - 3 - 4 237 96
FQ221041.1 - 3 - 4 237 96
FQ221174.1 - 3 - 4 235 96
FQ221230.1 - 3 - 4 310 96
FQ221308.1 - 3 - 4 239 96
FQ221518.1 - 3 - 4 237 96
FQ221677.1 - 3 - 4 235 96
FQ221825.1 - 3 - 4 237 96
FQ221877.1 - 3 - 4 237 96
FQ222252.1 - 3 - 4 248 96
FQ222272.1 - 3 - 4 237 96
FQ222274.1 - 3 - 4 237 96
FQ222304.1 - 3 - 4 237 96
FQ222376.1 - 3 - 4 237 96
FQ222415.1 - 3 - 4 235 96
FQ222418.1 - 3 - 4 239 96
FQ222718.1 - 3 - 4 237 96
FQ222721.1 - 3 - 4 235 96
FQ222768.1 - 3 - 4 235 96
FQ222892.1 - 3 - 4 238 96
FQ222922.1 - 3 - 4 306 96
FQ223006.1 - 3 - 4 237 96
FQ223095.1 - 3 - 4 237 96
FQ223230.1 - 3 - 4 236 96
FQ223389.1 - 3 - 4 237 96
FQ223433.1 - 3 - 4 237 96
FQ223787.1 - 3 - 4 237 96
FQ223930.1 - 359 96
FQ224201.1 - 3 - 4 237 96
FQ224347.1 - 3 - 4 237 96
FQ224383.1 - 3 - 4 237 96
FQ228419.1 - 3 - 4 236 96
FQ228429.1 - 3 - 4 237 96
FQ228446.1 - 3 - 4 237 96
FQ228475.1 - 3 - 4 238 96
FQ228508.1 - 3 - 4 237 96
FQ228609.1 - 3 - 4 237 96
FQ228617.1 - 3 - 4 236 96
FQ229004.1 - 3 - 4 238 96
FQ229042.1 - 3 - 4 235 96
FQ229063.1 - 3 - 4 236 96
X77953.1 - 2 - 3 216 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps15a

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
positive regulation of cell proliferation
response to virus
positive regulation of cell cycle
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100050800.rn.1 TC0100006483.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100050797.rn.1 TC0100006483.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100050799.rn.1 TC0100006483.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0