This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00672740_m1
See other GPR83 GE Assays ›
Gene Symbol:
Gpr83
Gene Name:
G protein-coupled receptor 83
Gene Aliases:
Gir
Chromosome Location:
Chr.8: 13365583 - 13376071 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00672740_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080411.1 NP_536336.1 3 - 4 954 84
GenBank mRNA AY029071.1 - 3 - 4 954 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000112.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr83

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

neuropeptide signaling pathway
response to glucocorticoid
neuropeptide Y receptor activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P269457 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 715619 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 861780 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 688712 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 124528 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 836045 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162132 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 516150 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 79149 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 135074 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269457 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269457 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P269457 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269457 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 100858 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 58212 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10908074 - -
Affymetrix Rat Gene 1.0 ST Array 10908078 - -
Affymetrix Rat Gene 1.0 ST Array 10908073 - -
Affymetrix Rat Gene 1.0 ST Array 10908079 - -
Affymetrix Rat Gene 1.0 ST Array 10908077 - -
Affymetrix Rat Gene 1.0 ST Array 10908075 - -
Affymetrix Rat Gene 1.1 ST Array 10908072 - -
Rat Genome 230 2.0 Array 1368299_at - -
Rat Genome 230 2.0 Array 1368299_PM_at - -
Affymetrix HT Rat Focus Array 1368299_PM_at - -
Affymetrix HT Rat Focus Array 1368299_at - -
Affymetrix Rat Clariom S Assay PSR0800000684.rn.1 TC0800000112.rn.1 RTA TC
Rat Expression Set 230 1368299_PM_at - -
Rat Expression Set 230 1368299_at - -
Affymetrix Rat Gene 1.0 ST Array 10908072 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20758634 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0