This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00583993_s1
See other GPR182 GE Assays ›
Gene Symbol:
Gpr182
Gene Name:
G protein-coupled receptor 182
Gene Aliases:
Admr
Chromosome Location:
Chr.7: 71044786 - 71048694 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00583993_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr182
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053302.3 NP_445754.2 2 903 84
XM_008765379.1 XP_008763601.1 2 1217 84
XM_008765380.1 XP_008763602.1 1 1267 84
GenBank mRNA BC129072.1 - 2 903 7096756 84
S79811.1 - 2 981 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700002929.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr182

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
G-protein coupled receptor signaling pathway
adrenomedullin receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12757 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 229605 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 944844 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 536287 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 146302 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12757 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12757 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12757 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12757 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700304474 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 36159 - -
Agilent Rat cDNA Microarray (G4105A) 700304474 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10903181 - -
Affymetrix Rat Gene 1.0 ST Array 10903178 - -
Affymetrix Rat Gene 1.0 ST Array 10903180 - -
Affymetrix Rat Gene 1.1 ST Array 10903177 - -
Rat Genome 230 2.0 Array 1369870_at - -
Rat Genome 230 2.0 Array 1369870_PM_at - -
Rat Genome 230 2.0 Array 1392664_at - -
Rat Genome 230 2.0 Array 1392664_PM_at - -
Affymetrix HT Rat Focus Array 1392664_at - -
Affymetrix HT Rat Focus Array 1369870_PM_at - -
Affymetrix HT Rat Focus Array 1392664_PM_at - -
Affymetrix HT Rat Focus Array 1369870_at - -
Affymetrix Rat Clariom S Assay PSR0700022789.rn.1 TC0700002929.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700022790.rn.1 TC0700002929.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700022792.rn.1 TC0700002929.rn.1 RTA TC
Rat Expression Set 230 1369870_PM_at - -
Rat Expression Set 230 1369870_at - -
Rat Expression Set 230 1392664_PM_at - -
Rat Expression Set 230 1392664_at - -
Affymetrix Rat Gene 1.0 ST Array 10903177 - -
Rat Genome U34 Set L04672_s_at - -
Rat Genome U34 Set rc_AA997122_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21474387 - -
ABI Rat Genome Survey Microarray 21540726 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371872 - -
RatRef-12 Expression BeadChip ILMN_57400 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0