This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00569139_m1
See other EDNRB GE Assays ›
Gene Symbol:
Ednrb
Gene Name:
endothelin receptor type B
Gene Aliases:
Ednra, Etb
Chromosome Location:
Chr.15: 88004775 - 88036354 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00569139_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ednrb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017333.1 NP_059029.1 2 - 3 741 79
XM_006252431.2 XP_006252493.1 2 - 3 1022 79
GenBank mRNA S65355.1 - 2 - 3 741 79
X57764.1 - 1 - 2 690 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500002296.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ednrb

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
regulation of pH
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
peripheral nervous system development
posterior midgut development
aging
regulation of blood pressure
positive regulation of cell proliferation
negative regulation of neuron maturation
response to organic cyclic compound
vein smooth muscle contraction
sensory perception of pain
cGMP-mediated signaling
melanocyte differentiation
regulation of fever generation
negative regulation of cellular protein metabolic process
response to lipopolysaccharide
enteric smooth muscle cell differentiation
positive regulation of urine volume
positive regulation of renal sodium excretion
epithelial fluid transport
vasoconstriction
vasodilation
negative regulation of apoptotic process
pigmentation
developmental pigmentation
macrophage chemotaxis
response to pain
enteric nervous system development
regulation of epithelial cell proliferation
regulation of sensory perception of pain
positive regulation of penile erection
cellular response to lipopolysaccharide
endothelin receptor signaling pathway
receptor activity
G-protein coupled receptor activity
endothelin receptor activity
peptide hormone binding
type 1 angiotensin receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500015540.rn.1 TC1500002296.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0