This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07224703_g1
See other ZNF577 GE Assays ›
Gene Symbol:
ZNF577
Gene Name:
zinc finger protein 577
Gene Aliases:
-
Chromosome Location:
Chr.19: 51614856 - 51642259 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07224703_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF577
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028839365.1 XP_028695198.1 400 86
XM_028839366.1 XP_028695199.1 401 86
XM_028839367.1 XP_028695200.1 352 86
GenBank mRNA JU336772.1 - 339 86
JU477532.1 - 337 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028839365.1, XM_028839366.1, XM_028839367.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0