This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04418391_s1
See other MIR199A-2 GE Assays ›
Gene Symbol:
MIR199A-2
Gene Name:
microRNA mir-199a-2
Gene Aliases:
mir-199a-2, mml-mir-199a-2
Chromosome Location:
Chr.19: 10581832 - 10581902 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04418391_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR199A-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_032443.1 38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0